Assay Details
| Gene Symbol: Dync1li1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146229.2 | NP_666341.1 | 8 - 9 | 1132 | 144 | |
| GenBank mRNA | AK159702.1 | - | 1174 | 144 | ||
| AK159983.1 | - | 121 | 144 | |||
| AK205150.1 | - | 2 - 3 | 193 | 144 | ||
| AK217382.1 | - | 2 - 3 | 144 | 144 | ||
| BC023347.1 | - | 8 - 9 | 1132 | 5040044 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dync1li1
Genomic Map