| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700020052.mm.1 |
NM_152810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020052.mm.1 |
NM_152810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020052.mm.1 |
TC1700002145.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020054.mm.1 |
NM_152810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020054.mm.1 |
NM_152810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020054.mm.1 |
TC1700002145.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020053.mm.1 |
NM_152810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020053.mm.1 |
NM_152810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020053.mm.1 |
TC1700002145.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020051.mm.1 |
NM_152810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020051.mm.1 |
NM_152810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020051.mm.1 |
TC1700002145.mm.1 |
MTA TC |