Assay Details
| Gene Symbol: Dnm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025947.2 | NP_001021118.1 | 10 - 11 | 1363 | 115 | |
| NM_001276340.1 | NP_001263269.1 | 10 - 11 | 1381 | 115 | ||
| NM_001276341.1 | NP_001263270.1 | 9 - 10 | 1125 | 115 | ||
| NM_152816.3 | NP_690029.2 | 11 - 12 | 1402 | 115 | ||
| NR_075074.1 | 10 - 11 | 1172 | 115 | |||
| GenBank mRNA | AB079133.1 | - | 10 - 11 | 1215 | 115 | |
| AK018195.1 | - | 11 - 12 | 1342 | 115 | ||
| AK051443.1 | - | 10 - 11 | 1292 | 115 | ||
| AK080871.1 | - | 10 - 11 | 1283 | 115 | ||
| AK154123.1 | - | 10 - 11 | 1132 | 115 | ||
| BC040777.1 | - | 9 - 10 | 1095 | 4948368 | 115 | |
| BC079635.1 | - | 10 - 11 | 1212 | 6400313 | 115 | |
| BC085158.1 | - | 11 - 12 | 1307 | 115 | ||
| BQ550714.1 | - | 2 - 3 | 299 | 115 | ||
| CD559921.1 | - | 2 - 3 | 225 | 115 | ||
| CK391623.1 | - | 2 - 3 | 150 | 115 | ||
| DV652251.1 | - | 3 - 4 | 389 | 115 | ||
More Information
Set Membership: |
|

Dnm1l
Genomic Map