Assay Details
| Gene Symbol: Dnm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025947.2 | NP_001021118.1 | 16 - 17 | 2051 | 147 | |
| NM_001276340.1 | NP_001263269.1 | 17 - 18 | 2102 | 147 | ||
| NM_001276341.1 | NP_001263270.1 | 16 - 17 | 1846 | 147 | ||
| NM_152816.3 | NP_690029.2 | 17 - 18 | 2090 | 147 | ||
| NR_075074.1 | 18 - 19 | 1971 | 147 | |||
| GenBank mRNA | AB079133.1 | - | 17 - 18 | 1936 | 147 | |
| AK018195.1 | - | 17 - 18 | 2030 | 147 | ||
| AK051443.1 | - | 16 - 17 | 1980 | 147 | ||
| AK080871.1 | - | 16 - 17 | 1971 | 147 | ||
| AK135420.1 | - | 2 - 3 | 958 | 147 | ||
| AK154123.1 | - | 18 - 19 | 1931 | 147 | ||
| BC040777.1 | - | 16 - 17 | 1816 | 4948368 | 147 | |
| BC079635.1 | - | 16 - 17 | 1900 | 6400313 | 147 | |
| BC085158.1 | - | 17 - 18 | 1995 | 148 | ||
| CK781757.1 | - | 2 - 3 | 216 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnm1l
Genomic Map