Assay Details
| Gene Symbol: Lhx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008499.5 | NP_032525.1 | 2 - 3 | 792 | 113 | |
| GenBank mRNA | AK033927.1 | - | 2 - 3 | 779 | 113 | |
| AK048778.1 | - | 2 - 3 | 781 | 113 | ||
| AK140208.1 | - | 2 - 3 | 793 | 113 | ||
| BC057585.1 | - | 2 - 3 | 680 | 6414270 | 113 | |
| U61155.1 | - | 2 - 3 | 908 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lhx5
Genomic Map