Assay Details
| Gene Symbol: Lta4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008517.2 | NP_032543.2 | 3 - 4 | 524 | 86 | |
| GenBank mRNA | AK134931.1 | - | 3 - 4 | 510 | 86 | |
| BC021417.1 | - | 3 - 4 | 518 | 5004113 | 86 | |
| M63848.1 | - | 490 | 86 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Lta4h
Genomic Map