Assay Details
| Gene Symbol: Mapt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038609.2 | NP_001033698.1 | 10 - 11 | 1149 | 82 | |
| NM_001285454.1 | NP_001272383.1 | 7 - 8 | 882 | 82 | ||
| NM_001285455.1 | NP_001272384.1 | 7 - 8 | 909 | 82 | ||
| NM_001285456.1 | NP_001272385.1 | 7 - 8 | 882 | 82 | ||
| NM_010838.4 | NP_034968.3 | 8 - 9 | 975 | 82 | ||
| GenBank mRNA | AF483518.1 | - | 7 - 8 | 790 | 82 | |
| AF483519.1 | - | 7 - 8 | 790 | 82 | ||
| AK078167.1 | - | 8 - 9 | 947 | 82 | ||
| AK147636.1 | - | 10 - 11 | 1110 | 82 | ||
| BC014748.1 | - | 8 - 9 | 913 | 4504850 | 82 | |
| M18775.1 | - | 7 - 8 | 883 | 82 | ||
| M18776.1 | - | 7 - 8 | 883 | 82 | ||
| M93266.1 | - | 3 - 4 | 294 | 82 | ||
| U12914.1 | - | 10 - 11 | 1033 | 82 | ||
| U12915.1 | - | 8 - 9 | 859 | 82 | ||
| U12916.1 | - | 7 - 8 | 793 | 82 | ||
| Z12133.1 | - | 3 - 4 | 294 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapt
Genomic Map