Assay Details
| Gene Symbol: Lgmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011175.2 | NP_035305.1 | 1 - 2 | 65 | 67 | |
| GenBank mRNA | AF044266.1 | - | 1 - 2 | 64 | 67 | |
| AJ000990.1 | - | 1 - 2 | 65 | 67 | ||
| AK149666.1 | - | 1 - 2 | 120 | 67 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF044266.1,AJ000990.1,AK149666.1,NM_011175.2 transcripts.
Set Membership: |
|

Lgmn
Genomic Map