Assay Details
| Gene Symbol: Lgmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011175.2 | NP_035305.1 | 10 - 11 | 914 | 75 | |
| GenBank mRNA | AF044266.1 | - | 10 - 11 | 913 | 75 | |
| AJ000990.1 | - | 10 - 11 | 914 | 75 | ||
| AK148104.1 | - | 9 - 10 | 863 | 75 | ||
| BC132515.1 | - | 9 - 10 | 861 | 40130792 | 75 | |
| BC132519.1 | - | 9 - 10 | 861 | 40130796 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgmn
Genomic Map