Assay Details
| Gene Symbol: Madcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013591.2 | NP_038619.2 | 3 - 4 | 688 | 84 | |
| GenBank mRNA | AK137028.1 | - | 3 - 4 | 689 | 84 | |
| L21203.1 | - | 3 - 4 | 685 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Madcam1
Genomic Map