Assay Details
| Gene Symbol: Madcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013591.2 | NP_038619.2 | 4 - 5 | 1114 | 120 | |
| GenBank mRNA | AK137028.1 | - | 4 - 5 | 1115 | 120 | |
| L21203.1 | - | 4 - 5 | 1111 | 120 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020326 | L21203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020326 | AK137028.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020326 | AK156096.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020326 | BC024372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020326 | NM_013591.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147803 | BE632565.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147803 | L21203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147803 | AK137028.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147803 | NM_013591.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347874 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125781 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147803 | BE632565.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147803 | L21203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147803 | AK137028.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147803 | NM_013591.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020326 | L21203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020326 | AK137028.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020326 | AK156096.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020326 | BC024372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020326 | NM_013591.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517745 | BC118056.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517745 | L21203.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517745 | S80258.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517745 | AK137028.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517745 | AK156096.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278109 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125781 | - | - |
| Whole Mouse Genome G4122A | A_51_P517745 | BC118056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517745 | L21203.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517745 | S80258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517745 | AK137028.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517745 | AK156096.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140287 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125781 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08745160 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08589153 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08745183 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08745165 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08745164 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08589159 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08745174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08263420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08745166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08205969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08589164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08517479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278094 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10364450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364445 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425253_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10364444 | - | - |
| Mouse Genome 430 2.0 Array | 1425253_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425253_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17123_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17123_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000005838.mm.1 | NM_013591.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005838.mm.1 | NM_013591.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005838.mm.1 | TC1000000808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005835.mm.1 | NM_013591.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005835.mm.1 | NM_013591.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005835.mm.1 | TC1000000808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005839.mm.1 | NM_013591.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005839.mm.1 | NM_013591.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005839.mm.1 | TC1000000808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005840.mm.1 | NM_013591.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005840.mm.1 | NM_013591.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005840.mm.1 | TC1000000808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005836.mm.1 | NM_013591.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005836.mm.1 | NM_013591.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005836.mm.1 | TC1000000808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005837.mm.1 | NM_013591.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005837.mm.1 | NM_013591.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005837.mm.1 | TC1000000808.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425253_PM_a_at | - | - |
| Mouse Expression Set 430 | 1425253_a_at | - | - |
| Murine Genome U74v2 Set | 102679_PM_at | - | - |
| Murine Genome U74v2 Set | 102680_PM_g_at | - | - |
| Murine Genome U74v2 Set | 102679_at | - | - |
| Murine Genome U74v2 Set | 102680_g_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425253_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425253_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425253_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425253_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1580010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001580010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2726874 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2726874 | - | - |
| mouse-6 v1.1 (Illumina) | scl38715.5.1_65-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1580010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38715.5.1_65-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2865448 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2865448 | - | - |

Madcam1
Genomic Map