Assay Details
| Gene Symbol: Mbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013594.2 | NP_038622.2 | 7 - 8 | 912 | 63 | |
| GenBank mRNA | AF072240.1 | - | 8 - 9 | 891 | 63 | |
| AK004624.2 | - | 7 - 8 | 913 | 63 | ||
| AK032535.1 | - | 4 - 5 | 410 | 63 | ||
| AK156401.1 | - | 8 - 9 | 903 | 63 | ||
| AK162858.1 | - | 7 - 8 | 912 | 63 | ||
| AK166042.1 | - | 3 - 4 | 232 | 63 | ||
| AK193128.1 | - | 3 - 4 | 208 | 63 | ||
| BC069837.1 | - | 7 - 8 | 928 | 30435760 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mbd1
Genomic Map