Assay Details
| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020267.2 | NP_064663.2 | 3 - 4 | 1205 | 58 | |
| GenBank mRNA | AJ249129.1 | - | 3 - 4 | 1154 | 58 | |
| AJ278191.1 | - | 3 - 4 | 1213 | 58 | ||
| BC039979.1 | - | 3 - 4 | 631 | 58 | ||
| BC045602.1 | - | 3 - 4 | 620 | 58 | ||
| BC098369.1 | - | 3 - 4 | 1174 | 6830438 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim44
Genomic Map