Assay Details
| Gene Symbol: Bcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020486.2 | NP_065232.1 | 3 - 4 | 476 | 91 | |
| GenBank mRNA | AB035511.1 | - | 3 - 4 | 437 | 91 | |
| AF109160.1 | - | 3 - 4 | 476 | 91 | ||
| AF221507.1 | - | 3 - 4 | 426 | 91 | ||
| AF346663.1 | - | 3 - 4 | 414 | 91 | ||
| AK040564.1 | - | 3 - 4 | 460 | 91 | ||
| AK054001.1 | - | 3 - 4 | 457 | 91 | ||
| AK075704.1 | - | 3 - 4 | 441 | 91 | ||
| BC004826.1 | - | 3 - 4 | 460 | 2616148 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcam
Genomic Map