Assay Details
| Gene Symbol: Bcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020486.2 | NP_065232.1 | 9 - 10 | 1235 | 82 | |
| GenBank mRNA | AF109160.1 | - | 9 - 10 | 1235 | 82 | |
| AF221507.1 | - | 9 - 10 | 1185 | 82 | ||
| AF346663.1 | - | 9 - 10 | 1173 | 82 | ||
| AK075704.1 | - | 9 - 10 | 1200 | 82 | ||
| AK158439.1 | - | 9 - 10 | 1201 | 82 | ||
| BC004826.1 | - | 9 - 10 | 1219 | 2616148 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcam
Genomic Map