Assay Details
| Gene Symbol: Ly6i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020498.2 | NP_065244.2 | 2 - 3 | 558 | 80 | |
| GenBank mRNA | AF232024.1 | - | 2 - 3 | 480 | 80 | |
| BC125350.1 | - | 2 - 3 | 558 | 40129865 | 80 | |
| BC132052.1 | - | 2 - 3 | 558 | 40130329 | 80 | |
| BC145087.1 | - | 3 - 4 | 300 | 9053635 | 80 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087118 | BC145087.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087118 | BC125350.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087118 | BC132052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087118 | NM_020498.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087118 | AF232024.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | AF065934.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | AF065935.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | AF065936.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | AF065937.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | AF065938.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | AK012356.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | BC027520.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | NM_011331.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | U50712.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | U66670.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | XM_001480891.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | XM_001480892.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913289 | XM_001480893.1 | refseq_mrna_mouse_XM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138474 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087118 | BC145087.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087118 | BC125350.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087118 | BC132052.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087118 | NM_020498.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087118 | AF232024.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30340621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30340617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30340634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30340606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30340594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30340599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30340635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30340588 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138474 | - | - |
| Whole Mouse Genome G4122A | A_51_P420918 | AF232024.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121400 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108967 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138474 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12366484 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12366475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30340566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12305082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12186373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12366474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11948177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12366482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12186369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30340568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10429561 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429562 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425890_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10429560 | - | - |
| Mouse Genome 430 2.0 Array | 1425890_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425890_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57248_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57248_at | - | - |
| Mouse Expression Set 430 | 1425890_PM_at | - | - |
| Mouse Expression Set 430 | 1425890_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425890_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425890_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425890_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425890_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 518342 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2810647 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002810647 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2741556 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2741556 | - | - |
| mouse-6 v1.1 (Illumina) | scl47071.3.1_38-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2810647 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47071.3.1_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2810647 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2741556 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2741556 | - | - |

Ly6i
Genomic Map