Assay Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190984.1 | NP_001177913.1 | 2 - 3 | 160 | 64 | |
| NM_001190985.1 | NP_001177914.1 | 1 - 2 | 378 | 64 | ||
| NM_021295.3 | NP_067270.1 | 2 - 3 | 194 | 64 | ||
| GenBank mRNA | AJ294535.1 | - | 2 - 3 | 187 | 64 | |
| AK146120.1 | - | 1 - 2 | 370 | 64 | ||
| AK182687.1 | - | 1 - 2 | 88 | 64 | ||
| AK186065.1 | - | 1 - 2 | 65 | |||
| AK210427.1 | - | 1 - 2 | 88 | 64 | ||
| BC058560.1 | - | 2 - 3 | 160 | 6315377 | 64 | |
| Y11550.1 | - | 2 - 3 | 159 | 64 | ||
| Y16518.1 | - | 1 - 2 | 338 | 64 | ||
More Information
Set Membership: |
|

Lancl1
Genomic Map