Assay Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190984.1 | NP_001177913.1 | 8 - 9 | 1130 | 73 | |
| NM_001190985.1 | NP_001177914.1 | 7 - 8 | 1348 | 73 | ||
| NM_021295.3 | NP_067270.1 | 8 - 9 | 1164 | 73 | ||
| GenBank mRNA | AJ294535.1 | - | 8 - 9 | 1157 | 73 | |
| AK146120.1 | - | 7 - 8 | 1339 | 73 | ||
| BC038649.1 | - | 5 - 6 | 580 | 73 | ||
| BC058560.1 | - | 8 - 9 | 1130 | 6315377 | 73 | |
| Y11550.1 | - | 8 - 9 | 1129 | 73 | ||
| Y16518.1 | - | 7 - 8 | 1308 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lancl1
Genomic Map