Assay Details
| Gene Symbol: Mapk8ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021921.3 | NP_068740.3 | 7 - 8 | 2347 | 109 | |
| GenBank mRNA | AF220195.1 | - | 7 - 8 | 2066 | 109 | |
| AF310135.1 | - | 7 - 8 | 2128 | 109 | ||
| AK014339.1 | - | 7 - 8 | 2051 | 109 | ||
| AK158968.1 | - | 7 - 8 | 2201 | 109 | ||
| BC029704.1 | - | 7 - 8 | 2126 | 5359447 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk8ip2
Genomic Map