Assay Details
| Gene Symbol: Mark3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022801.4 | NP_073712.2 | 15 - 16 | 2388 | 90 | |
| GenBank mRNA | AF240783.1 | - | 15 - 16 | 1933 | 90 | |
| AK166157.1 | - | 15 - 16 | 2377 | 90 | ||
| BC026445.1 | - | 8 - 9 | 1293 | 4459439 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mark3
Genomic Map