Assay Details
| Gene Symbol: Mcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023061.2 | NP_075548.2 | 5 - 6 | 590 | 96 | |
| GenBank mRNA | AB035508.1 | - | 5 - 6 | 577 | 96 | |
| AB035509.1 | - | 5 - 6 | 577 | 96 | ||
| BC026985.1 | - | 5 - 6 | 577 | 5372497 | 96 | |
| BQ178999.1 | - | 3 - 4 | 188 | 96 | ||
| CN699979.1 | - | 5 - 6 | 536 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcam
Genomic Map