Assay Details
| Gene Symbol: Mcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023061.2 | NP_075548.2 | 7 - 8 | 899 | 66 | |
| GenBank mRNA | AB035508.1 | - | 7 - 8 | 886 | 66 | |
| AB035509.1 | - | 7 - 8 | 886 | 66 | ||
| BC026985.1 | - | 7 - 8 | 886 | 5372497 | 66 | |
| BQ178999.1 | - | 5 - 6 | 497 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcam
Genomic Map