Assay Details
| Gene Symbol: Mccc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023644.4 | NP_076133.3 | 1 - 2 | 196 | 132 | |
| GenBank mRNA | AF310338.1 | - | 1 - 2 | 117 | 132 | |
| AK007782.1 | - | 1 - 2 | 149 | 132 | ||
| AK031072.1 | - | 1 - 2 | 157 | 132 | ||
| AK032203.1 | - | 1 - 2 | 128 | 132 | ||
| AK168589.1 | - | 1 - 2 | 85 | 132 | ||
| AK178493.1 | - | 1 - 2 | 50 | 132 | ||
| BC021382.1 | - | 1 - 2 | 160 | 5042993 | 132 | |
| BY139695.1 | - | 1 - 2 | 197 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mccc1
Genomic Map