Assay Details
| Gene Symbol: Mccc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023644.4 | NP_076133.3 | 14 - 15 | 1787 | 70 | |
| GenBank mRNA | AF310338.1 | - | 14 - 15 | 1708 | 70 | |
| AK007782.1 | - | 14 - 15 | 1740 | 70 | ||
| AK031072.1 | - | 14 - 15 | 1748 | 70 | ||
| AK032203.1 | - | 14 - 15 | 1730 | 70 | ||
| AK168589.1 | - | 14 - 15 | 1676 | 70 | ||
| BC021382.1 | - | 14 - 15 | 1751 | 5042993 | 70 | |
More Information
Set Membership: |
|

Mccc1
Genomic Map