Assay Details
| Gene Symbol: Pdzk1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164557.1 | NP_001158029.1 | 1 - 2 | 508 | 71 | |
| NM_001164558.1 | NP_001158030.1 | 2 - 3 | 205 | 71 | ||
| NM_026018.3 | NP_080294.1 | 2 - 3 | 276 | 71 | ||
| GenBank mRNA | AB009296.1 | - | 1 - 2 | 86 | 71 | |
| AK007936.1 | - | 1 - 2 | 494 | 71 | ||
| AK008253.1 | - | 2 - 3 | 262 | 71 | ||
| AK012305.1 | - | 2 - 3 | 223 | 71 | ||
| BC013542.1 | - | 1 - 2 | 113 | 4241868 | 71 | |
| DV057288.1 | - | 2 - 3 | 222 | 71 | ||
| DV058382.1 | - | 1 - 2 | 197 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdzk1ip1
Genomic Map