| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700030983.mm.1 |
NM_030705.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030983.mm.1 |
NM_030705.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030983.mm.1 |
TC0700003647.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030982.mm.1 |
NM_030705.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030982.mm.1 |
NM_030705.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030982.mm.1 |
TC0700003647.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030981.mm.1 |
NM_030705.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030981.mm.1 |
NM_030705.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030981.mm.1 |
TC0700003647.mm.1 |
MTA TC |