Assay Details
| Gene Symbol: Lbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133815.2 | NP_598576.2 | 4 - 5 | 607 | 91 | |
| GenBank mRNA | AK028426.1 | - | 4 - 5 | 584 | 91 | |
| AK030606.1 | - | 4 - 5 | 561 | 91 | ||
| AK033459.1 | - | 4 - 5 | 559 | 91 | ||
| AK088179.1 | - | 4 - 5 | 560 | 91 | ||
| AK151798.1 | - | 4 - 5 | 574 | 91 | ||
| AK161762.1 | - | 4 - 5 | 585 | 91 | ||
| AK166850.1 | - | 4 - 5 | 547 | 91 | ||
| AK167157.1 | - | 4 - 5 | 552 | 91 | ||
| AK199017.1 | - | 1 - 2 | 94 | 91 | ||
| BC021516.1 | - | 4 - 5 | 548 | 5352337 | 91 | |
| BC025583.1 | - | 3 - 4 | 357 | 91 | ||
| BC042522.1 | - | 4 - 5 | 567 | 4216442 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lbr
Genomic Map