Assay Details
| Gene Symbol: Lbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133815.2 | NP_598576.2 | 10 - 11 | 1474 | 102 | |
| GenBank mRNA | AK028426.1 | - | 10 - 11 | 1451 | 102 | |
| AK030606.1 | - | 10 - 11 | 1428 | 102 | ||
| AK033459.1 | - | 10 - 11 | 1426 | 102 | ||
| AK088179.1 | - | 10 - 11 | 1427 | 102 | ||
| AK089042.1 | - | 1 - 2 | 188 | 102 | ||
| AK151798.1 | - | 10 - 11 | 1441 | 102 | ||
| AK161762.1 | - | 10 - 11 | 1452 | 102 | ||
| AK166850.1 | - | 10 - 11 | 1414 | 102 | ||
| AK167157.1 | - | 10 - 11 | 1419 | 102 | ||
| BC010261.1 | - | 3 - 4 | 424 | 102 | ||
| BC014835.1 | - | 2 - 3 | 217 | 102 | ||
| BC021516.1 | - | 10 - 11 | 1415 | 5352337 | 102 | |
| BC025583.1 | - | 9 - 10 | 1223 | 102 | ||
| BC029171.1 | - | 3 - 4 | 421 | 102 | ||
| BC042522.1 | - | 10 - 11 | 1434 | 4216442 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lbr
Genomic Map