Assay Details
| Gene Symbol: Slc25a38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144793.1 | NP_659042.1 | 4 - 5 | 553 | 74 | |
| GenBank mRNA | AK078385.1 | - | 4 - 5 | 569 | 74 | |
| BC010801.1 | - | 4 - 5 | 553 | 4235245 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a38
Genomic Map