Assay Details
| Gene Symbol: Slc25a38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144793.1 | NP_659042.1 | 6 - 7 | 907 | 64 | |
| GenBank mRNA | AK078385.1 | - | 6 - 7 | 923 | 64 | |
| BC010801.1 | - | 6 - 7 | 907 | 4235245 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876103 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876102 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876104 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876099 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964068 | NM_144793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964068 | AK078385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964068 | BC010801.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249900 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576051 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700537 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24688 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121417 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144043 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964068 | NM_144793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964068 | AK078385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964068 | BC010801.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26876170 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144043 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02045579 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06948581 | - | - |
| Whole Mouse Genome G4122A | A_51_P427425 | NM_144793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427425 | AK078385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427425 | BC010801.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107164 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06948581 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144043 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07520217 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07520215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876104 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876102 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26876103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26876107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08017506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07482883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07814400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07482885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07814401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07814404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07520214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08194869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08193404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07482887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07942086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07831514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26876133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07975893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07503906 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10590246 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590251 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590249 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590247 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590250 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590248 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424418_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10590245 | - | - |
| Mouse Genome 430 2.0 Array | 1424418_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424418_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208638_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208638_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900013327.mm.1 | NM_144793.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013327.mm.1 | NM_144793.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013327.mm.1 | TC0900001594.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013325.mm.1 | NM_144793.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013325.mm.1 | NM_144793.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013325.mm.1 | TC0900001594.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424418_at | - | - |
| Mouse Expression Set 430 | 1424418_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424418_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424418_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424418_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424418_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 624796 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2510039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760273 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2510039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002510039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760273 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259600 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256420 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256420 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259600 | - | - |
| mouse-6 v1.1 (Illumina) | scl36342.8.1_2-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003479.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760273 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2510039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003479.1_30-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36342.8.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2510039 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2946905 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2946901 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256420 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2946901 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2946905 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256420 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36342.8.1_2-S | - | - |

Slc25a38
Genomic Map