| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700013455.mm.1 |
NM_144809.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013455.mm.1 |
NM_144809.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013455.mm.1 |
TC1700001501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013464.mm.1 |
NM_144809.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013464.mm.1 |
NM_144809.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013464.mm.1 |
TC1700001501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013461.mm.1 |
NM_144809.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013461.mm.1 |
NM_144809.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013461.mm.1 |
TC1700001501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013458.mm.1 |
TC1700001501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013460.mm.1 |
NM_144809.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013460.mm.1 |
NM_144809.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013460.mm.1 |
TC1700001501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013457.mm.1 |
NM_144809.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013457.mm.1 |
NM_144809.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013457.mm.1 |
TC1700001501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013463.mm.1 |
NM_144809.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013463.mm.1 |
NM_144809.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013463.mm.1 |
TC1700001501.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013467.mm.1 |
TC1700001501.mm.1 |
MTA TC |