Assay Details
| Gene Symbol: AI317395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144821.4 | NP_659070.2 | 1 - 2 | 377 | 57 | |
| GenBank mRNA | AK089961.1 | - | 1 - 2 | 380 | 57 | |
| BC018406.1 | - | 1 - 2 | 281 | 4196864 | 57 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK089961.1,BC018406.1,NM_144821.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27081964 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27081962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27081965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27081963 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436469 | AK089961.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436469 | NM_144821.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436469 | AK089961.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436469 | NM_144821.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515855 | AK019589.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515855 | XM_001474455.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515855 | XM_001479733.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257212 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477740 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704562 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 114734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161081 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164157 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125258 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436469 | AK089961.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436469 | NM_144821.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436469 | AK089961.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436469 | NM_144821.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436469 | AK089961.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436469 | NM_144821.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436469 | AK089961.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436469 | NM_144821.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27081946 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125258 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07035223 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164157 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07035242 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07035199 | - | - |
| Whole Mouse Genome G4122A | A_51_P436469 | AK089961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436469 | NM_144821.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436469 | AK089961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436469 | NM_144821.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137778 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104884 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164157 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07035223 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125258 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08414430 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08414421 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08414433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27081964 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27081962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27081965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27081963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27082006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27082013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27082008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27082004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27082002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27082010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27082000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27082005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27081880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27082013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08334896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08288541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08326597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27082006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08414420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27082009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27082000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08233876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08582915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27082010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27082007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08457789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08414433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08414436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08951445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08218289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27082008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08770111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27082004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08414409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27082002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27081915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08582910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10368717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368718 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425596_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436109_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368713 | - | - |
| Mouse Genome 430 2.0 Array | 1425596_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436109_at | - | - |
| Mouse Genome 430 2.0 Array | 1451760_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1451760_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1425596_at | - | - |
| Mouse Genome 430 2.0 Array | 1436109_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 215929_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 215929_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000014456.mm.1 | NM_144821.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014456.mm.1 | NM_144821.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014456.mm.1 | TC1000002035.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000014459.mm.1 | NM_144821.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014459.mm.1 | NM_144821.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014459.mm.1 | TC1000002035.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000014457.mm.1 | NM_144821.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014457.mm.1 | NM_144821.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014457.mm.1 | TC1000002035.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451760_PM_s_at | - | - |
| Mouse Expression Set 430 | 1425596_at | - | - |
| Mouse Expression Set 430 | 1425596_PM_at | - | - |
| Mouse Expression Set 430 | 1451760_s_at | - | - |
| Mouse Expression Set 430 | 1436109_PM_at | - | - |
| Mouse Expression Set 430 | 1436109_at | - | - |
| Murine Genome U74v2 Set | 169304_PM_at | - | - |
| Murine Genome U74v2 Set | 170836_at | - | - |
| Murine Genome U74v2 Set | 169304_at | - | - |
| Murine Genome U74v2 Set | 170836_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425596_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425596_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451760_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451760_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425596_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425596_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436109_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436109_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 510149 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 100630129 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6220072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006220072 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2594156 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2594156 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220558 | - | - |
| mouse-6 v1.1 (Illumina) | scl38034.5_53-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6220072 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100630129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38034.5_53-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6220072 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100630129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2987599 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220558 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2987599 | - | - |

AI317395
Genomic Map