Assay Details
| Gene Symbol: Acsl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033597.1 | NP_001028769.1 | 8 - 9 | 945 | 66 | |
| NM_001033598.1 | NP_001028770.1 | 8 - 9 | 914 | 66 | ||
| NM_001033599.1 | NP_001028771.1 | 8 - 9 | 1120 | 66 | ||
| NM_144823.4 | NP_659072.3 | 8 - 9 | 945 | 66 | ||
| GenBank mRNA | AB073974.1 | - | 7 - 8 | 789 | 66 | |
| AK032479.1 | - | 9 - 10 | 1204 | 66 | ||
| AK039380.1 | - | 8 - 9 | 1031 | 66 | ||
| AK147283.1 | - | 8 - 9 | 915 | 66 | ||
| AK147421.1 | - | 8 - 9 | 946 | 66 | ||
| AY167035.1 | - | 8 - 9 | 66 | |||
| AY786360.1 | - | 10 - 11 | 1463 | 66 | ||
| AY786361.1 | - | 8 - 9 | 895 | 66 | ||
| AY786362.1 | - | 8 - 9 | 1087 | 66 | ||
| AY786363.1 | - | 8 - 9 | 940 | 66 | ||
| BC016114.1 | - | 8 - 9 | 871 | 4481949 | 66 | |
| BC022959.1 | - | 8 - 9 | 945 | 5362660 | 66 | |
More Information
Set Membership: |
|

Acsl6
Genomic Map