Assay Details
| Gene Symbol: Acsl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033597.1 | NP_001028769.1 | 13 - 14 | 1417 | 114 | |
| NM_001033598.1 | NP_001028770.1 | 13 - 14 | 1386 | 114 | ||
| NM_001033599.1 | NP_001028771.1 | 13 - 14 | 1592 | 114 | ||
| NM_144823.4 | NP_659072.3 | 13 - 14 | 1417 | 114 | ||
| GenBank mRNA | AK122384.1 | - | 2 - 3 | 3183 | 114 | |
| AK147283.1 | - | 13 - 14 | 1387 | 114 | ||
| AK147421.1 | - | 13 - 14 | 1418 | 114 | ||
| AY167035.1 | - | 13 - 14 | 1387 | 114 | ||
| AY786360.1 | - | 15 - 16 | 1935 | 114 | ||
| AY786361.1 | - | 13 - 14 | 1367 | 114 | ||
| AY786362.1 | - | 13 - 14 | 1559 | 114 | ||
| AY786363.1 | - | 13 - 14 | 1412 | 114 | ||
| BC016114.1 | - | 13 - 14 | 1343 | 4481949 | 114 | |
| BC022959.1 | - | 13 - 14 | 1417 | 5362660 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsl6
Genomic Map