Assay Details
| Gene Symbol: Wrap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144824.2 | NP_659073.1 | 6 - 7 | 1183 | 106 | |
| GenBank mRNA | AK077570.1 | - | 6 - 7 | 1087 | 106 | |
| AK145219.1 | - | 6 - 7 | 1110 | 106 | ||
| AK169271.1 | - | 5 - 6 | 1196 | 106 | ||
| BC021790.1 | - | 6 - 7 | 1183 | 4220923 | 106 | |
| BC069868.1 | - | 7 - 8 | 1227 | 5101220 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wrap53
Genomic Map