Assay Details
| Gene Symbol: Wrap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144824.2 | NP_659073.1 | 8 - 9 | 1488 | 62 | |
| GenBank mRNA | AK077570.1 | - | 8 - 9 | 1392 | 62 | |
| AK145219.1 | - | 8 - 9 | 1415 | 62 | ||
| AK169271.1 | - | 7 - 8 | 1501 | 62 | ||
| BC021790.1 | - | 8 - 9 | 1488 | 4220923 | 62 | |
| BC069868.1 | - | 9 - 10 | 1532 | 5101220 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wrap53
Genomic Map