Assay Details
| Gene Symbol: Serpina10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301404.1 | NP_001288333.1 | 3 - 4 | 943 | 66 | |
| NM_144834.4 | NP_659083.2 | 3 - 4 | 1105 | 66 | ||
| GenBank mRNA | AK050552.1 | - | 1082 | 66 | ||
| AY324633.1 | - | 3 - 4 | 1105 | 66 | ||
| BC018416.1 | - | 3 - 4 | 920 | 4196269 | 66 | |
| BC025821.1 | - | 2 - 3 | 1067 | 5100400 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serpina10
Genomic Map