Assay Details
| Gene Symbol: Mtmr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144843.4 | NP_659092.1 | 11 - 12 | 1479 | 66 | |
| GenBank mRNA | AB517623.1 | - | 12 - 13 | 1460 | 66 | |
| AK076218.1 | - | 11 - 12 | 1456 | 66 | ||
| AK154859.1 | - | 11 - 12 | 1399 | 66 | ||
| BC020019.1 | - | 11 - 12 | 1413 | 3596732 | 66 | |
| BC022979.1 | - | 1 - 2 | 520 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr6
Genomic Map