Assay Details
| Gene Symbol: Senp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144851.5 | NP_659100.1 | 14 - 15 | 1923 | 107 | |
| GenBank mRNA | AK028565.1 | - | 14 - 15 | 1665 | 107 | |
| AK028762.1 | - | 6 - 7 | 2897 | 107 | ||
| AK035723.1 | - | 15 - 16 | 2106 | 107 | ||
| AK054521.1 | - | 15 - 16 | 2055 | 107 | ||
| AK088597.1 | - | 14 - 15 | 1667 | 107 | ||
| BC023129.1 | - | 13 - 14 | 1637 | 3595534 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Senp1
Genomic Map