| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500017534.mm.1 |
NM_144851.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017534.mm.1 |
NM_144851.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017534.mm.1 |
TC1500002165.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017537.mm.1 |
NM_144851.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017537.mm.1 |
NM_144851.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017537.mm.1 |
TC1500002165.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017538.mm.1 |
NM_144851.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017538.mm.1 |
NM_144851.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017538.mm.1 |
TC1500002165.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017535.mm.1 |
NM_144851.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017535.mm.1 |
NM_144851.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017535.mm.1 |
TC1500002165.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017540.mm.1 |
NM_144851.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017540.mm.1 |
NM_144851.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017540.mm.1 |
TC1500002165.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017536.mm.1 |
NM_144851.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017536.mm.1 |
NM_144851.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017536.mm.1 |
TC1500002165.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017541.mm.1 |
NM_144851.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017541.mm.1 |
NM_144851.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017541.mm.1 |
TC1500002165.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017539.mm.1 |
NM_144851.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017539.mm.1 |
NM_144851.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017539.mm.1 |
TC1500002165.mm.1 |
MTA TC |