Assay Details
| Gene Symbol: Etf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144866.3 | NP_659115.3 | 6 - 7 | 918 | 130 | |
| GenBank mRNA | AK049607.1 | - | 6 - 7 | 869 | 130 | |
| AK077559.1 | - | 6 - 7 | 867 | 130 | ||
| AK139933.1 | - | 6 - 7 | 881 | 131 | ||
| AK151710.1 | - | 6 - 7 | 819 | 130 | ||
| AK152428.1 | - | 6 - 7 | 819 | 130 | ||
| AK152952.1 | - | 6 - 7 | 819 | 130 | ||
| AK164020.1 | - | 6 - 7 | 869 | 130 | ||
| AK166740.1 | - | 6 - 7 | 887 | 130 | ||
| AK169345.1 | - | 6 - 7 | 891 | 130 | ||
| AK170221.1 | - | 6 - 7 | 861 | 130 | ||
| BC013717.1 | - | 6 - 7 | 845 | 3992883 | 130 | |
| D87691.1 | - | 3 - 4 | 433 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Etf1
Genomic Map