Assay Details
| Gene Symbol: Etf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144866.3 | NP_659115.3 | 9 - 10 | 1269 | 112 | |
| GenBank mRNA | AK049607.1 | - | 9 - 10 | 1220 | 112 | |
| AK077559.1 | - | 9 - 10 | 1219 | 112 | ||
| AK139933.1 | - | 9 - 10 | 1233 | 112 | ||
| AK151028.1 | - | 9 - 10 | 1169 | 112 | ||
| AK151710.1 | - | 9 - 10 | 1170 | 112 | ||
| AK152428.1 | - | 9 - 10 | 1170 | 112 | ||
| AK152952.1 | - | 9 - 10 | 1170 | 112 | ||
| AK164020.1 | - | 9 - 10 | 1220 | 112 | ||
| AK166740.1 | - | 9 - 10 | 1238 | 112 | ||
| AK169345.1 | - | 9 - 10 | 1242 | 112 | ||
| AK170221.1 | - | 9 - 10 | 1212 | 112 | ||
| BC013717.1 | - | 9 - 10 | 1196 | 3992883 | 112 | |
| D87691.1 | - | 6 - 7 | 784 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Etf1
Genomic Map