Assay Details
| Gene Symbol: Cox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144874.4 | NP_659123.2 | 7 - 8 | 1082 | 118 | |
| GenBank mRNA | AK048099.1 | - | 7 - 8 | 1064 | 118 | |
| AK051792.1 | - | 7 - 8 | 981 | 118 | ||
| AK141631.1 | - | 7 - 8 | 1014 | 118 | ||
| BC011509.1 | - | 7 - 8 | 1042 | 3981950 | 118 | |
| BC021498.1 | - | 7 - 8 | 1002 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox15
Genomic Map