Assay Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 1 - 2 | 304 | 52 | |
| GenBank mRNA | AK088000.1 | - | 1 - 2 | 265 | 52 | |
| AK137001.1 | - | 1 - 2 | 263 | 52 | ||
| AK159279.1 | - | 1 - 2 | 261 | 52 | ||
| AK159306.1 | - | 1 - 2 | 261 | 52 | ||
| BB863666.1 | - | 1 - 2 | 306 | 52 | ||
| BC021496.1 | - | 1 - 2 | 254 | 5346159 | 52 | |
| BC023794.1 | - | 2 - 3 | 199 | 5351943 | 52 | |
| BC023964.1 | - | 1 - 2 | 244 | 52 | ||
| BC025618.1 | - | 1 - 2 | 263 | 5320976 | 52 | |
| BC025627.1 | - | 1 - 2 | 254 | 5320513 | 52 | |
| BC025811.1 | - | 1 - 2 | 241 | 5096872 | 52 | |
| BC032187.1 | - | 1 - 2 | 304 | 5321268 | 52 | |
| BC033435.1 | - | 1 - 2 | 200 | 4950746 | 52 | |
| BC033471.1 | - | 1 - 2 | 244 | 4954119 | 52 | |
| BC042435.1 | - | 1 - 2 | 254 | 4951288 | 52 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK159279.1,BC021496.1,BC023794.1,BC025618.1,BC025627.1,BC025811.1,BC032187.1,BC033435.1,BC033471.1,BC042435.1,NM_144900.2 transcripts.
Set Membership: |
|

Atp1a1
Genomic Map