Assay Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 4 - 5 | 685 | 71 | |
| GenBank mRNA | AK088000.1 | - | 4 - 5 | 646 | 71 | |
| AK137001.1 | - | 4 - 5 | 644 | 71 | ||
| AK159279.1 | - | 4 - 5 | 642 | 71 | ||
| AK206258.1 | - | 1 - 2 | 184 | 71 | ||
| BC010319.1 | - | 2 - 3 | 270 | 71 | ||
| BC021496.1 | - | 4 - 5 | 635 | 5346159 | 71 | |
| BC023794.1 | - | 5 - 6 | 580 | 5351943 | 71 | |
| BC023964.1 | - | 4 - 5 | 625 | 71 | ||
| BC025618.1 | - | 4 - 5 | 644 | 5320976 | 71 | |
| BC025627.1 | - | 4 - 5 | 635 | 5320513 | 71 | |
| BC025811.1 | - | 4 - 5 | 622 | 5096872 | 71 | |
| BC032187.1 | - | 4 - 5 | 685 | 5321268 | 71 | |
| BC033435.1 | - | 4 - 5 | 581 | 4950746 | 71 | |
| BC033471.1 | - | 4 - 5 | 625 | 4954119 | 71 | |
| BC034177.1 | - | 3 - 4 | 390 | 71 | ||
| BC035352.1 | - | 3 - 4 | 355 | 71 | ||
| BC042435.1 | - | 4 - 5 | 635 | 4951288 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a1
Genomic Map