Assay Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 6 - 7 | 929 | 85 | |
| GenBank mRNA | AK088000.1 | - | 6 - 7 | 890 | 85 | |
| AK137001.1 | - | 6 - 7 | 888 | 85 | ||
| AK159279.1 | - | 6 - 7 | 886 | 85 | ||
| AK159306.1 | - | 6 - 7 | 884 | 85 | ||
| BC010319.1 | - | 4 - 5 | 514 | 85 | ||
| BC021496.1 | - | 6 - 7 | 879 | 5346159 | 85 | |
| BC023794.1 | - | 7 - 8 | 824 | 5351943 | 85 | |
| BC023964.1 | - | 6 - 7 | 869 | 85 | ||
| BC025618.1 | - | 6 - 7 | 888 | 5320976 | 85 | |
| BC025627.1 | - | 6 - 7 | 879 | 5320513 | 85 | |
| BC025811.1 | - | 6 - 7 | 866 | 5096872 | 85 | |
| BC032187.1 | - | 6 - 7 | 929 | 5321268 | 85 | |
| BC033435.1 | - | 6 - 7 | 825 | 4950746 | 85 | |
| BC033471.1 | - | 6 - 7 | 869 | 4954119 | 85 | |
| BC034177.1 | - | 5 - 6 | 634 | 85 | ||
| BC035352.1 | - | 5 - 6 | 599 | 85 | ||
| BC042435.1 | - | 6 - 7 | 879 | 4951288 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a1
Genomic Map