Assay Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 9 - 10 | 1519 | 59 | |
| GenBank mRNA | AK088000.1 | - | 9 - 10 | 1480 | 59 | |
| AK137001.1 | - | 9 - 10 | 1478 | 59 | ||
| AK150523.1 | - | 2 - 3 | 424 | 59 | ||
| AK159279.1 | - | 9 - 10 | 1476 | 59 | ||
| AK159306.1 | - | 9 - 10 | 1474 | 59 | ||
| BC010319.1 | - | 7 - 8 | 1104 | 59 | ||
| BC021496.1 | - | 9 - 10 | 1469 | 5346159 | 59 | |
| BC023794.1 | - | 10 - 11 | 1414 | 5351943 | 59 | |
| BC023964.1 | - | 9 - 10 | 1459 | 59 | ||
| BC025037.1 | - | 2 - 3 | 319 | 59 | ||
| BC025618.1 | - | 9 - 10 | 1478 | 5320976 | 59 | |
| BC025627.1 | - | 9 - 10 | 1469 | 5320513 | 59 | |
| BC025811.1 | - | 9 - 10 | 1456 | 5096872 | 59 | |
| BC032187.1 | - | 9 - 10 | 1519 | 5321268 | 59 | |
| BC033435.1 | - | 9 - 10 | 1415 | 4950746 | 59 | |
| BC033471.1 | - | 9 - 10 | 1459 | 4954119 | 59 | |
| BC034177.1 | - | 8 - 9 | 1224 | 59 | ||
| BC035352.1 | - | 8 - 9 | 1189 | 59 | ||
| BC042435.1 | - | 9 - 10 | 1469 | 4951288 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a1
Genomic Map