Assay Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 13 - 14 | 2129 | 71 | |
| GenBank mRNA | AK088000.1 | - | 13 - 14 | 2090 | 71 | |
| AK137001.1 | - | 13 - 14 | 2088 | 71 | ||
| AK150523.1 | - | 6 - 7 | 1034 | 71 | ||
| AK159279.1 | - | 13 - 14 | 2086 | 71 | ||
| AK159306.1 | - | 13 - 14 | 2084 | 71 | ||
| AK184412.1 | - | 1 - 2 | 186 | 71 | ||
| AK199691.1 | - | 1 - 2 | 188 | 71 | ||
| AK211215.1 | - | 1 - 2 | 186 | 71 | ||
| BC010319.1 | - | 11 - 12 | 1714 | 71 | ||
| BC021496.1 | - | 13 - 14 | 2079 | 5346159 | 71 | |
| BC023794.1 | - | 14 - 15 | 2024 | 5351943 | 71 | |
| BC023964.1 | - | 13 - 14 | 2070 | 71 | ||
| BC025037.1 | - | 6 - 7 | 929 | 71 | ||
| BC025618.1 | - | 13 - 14 | 2088 | 5320976 | 71 | |
| BC025627.1 | - | 13 - 14 | 2079 | 5320513 | 71 | |
| BC025811.1 | - | 13 - 14 | 2066 | 5096872 | 71 | |
| BC030326.1 | - | 2 - 3 | 426 | 71 | ||
| BC032187.1 | - | 13 - 14 | 2129 | 5321268 | 71 | |
| BC033435.1 | - | 13 - 14 | 2025 | 4950746 | 71 | |
| BC033471.1 | - | 13 - 14 | 2069 | 4954119 | 71 | |
| BC034177.1 | - | 12 - 13 | 1834 | 71 | ||
| BC035352.1 | - | 12 - 13 | 1799 | 71 | ||
| BC042435.1 | - | 13 - 14 | 2079 | 4951288 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a1
Genomic Map