Assay Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 18 - 19 | 2866 | 98 | |
| GenBank mRNA | AK088000.1 | - | 18 - 19 | 2827 | 98 | |
| AK137001.1 | - | 18 - 19 | 2826 | 98 | ||
| AK148989.1 | - | 5 - 6 | 724 | 98 | ||
| AK150523.1 | - | 11 - 12 | 1771 | 98 | ||
| AK159279.1 | - | 18 - 19 | 2823 | 98 | ||
| AK159306.1 | - | 18 - 19 | 2821 | 98 | ||
| AK189977.1 | - | 2 - 3 | 221 | 98 | ||
| AK190187.1 | - | 2 - 3 | 220 | 98 | ||
| AK191797.1 | - | 2 - 3 | 220 | 98 | ||
| AK192305.1 | - | 2 - 3 | 220 | 98 | ||
| AK192395.1 | - | 2 - 3 | 220 | 98 | ||
| AK192706.1 | - | 319 | 98 | |||
| AK193322.1 | - | 2 - 3 | 220 | 98 | ||
| AK195171.1 | - | 2 - 3 | 220 | 98 | ||
| AK196676.1 | - | 2 - 3 | 220 | 98 | ||
| AK197178.1 | - | 2 - 3 | 220 | 98 | ||
| AK197377.1 | - | 2 - 3 | 220 | 98 | ||
| AK197675.1 | - | 2 - 3 | 221 | 98 | ||
| AK199023.1 | - | 2 - 3 | 220 | 98 | ||
| AK201233.1 | - | 2 - 3 | 220 | 98 | ||
| AK202370.1 | - | 2 - 3 | 221 | 98 | ||
| AK202506.1 | - | 2 - 3 | 221 | 98 | ||
| AK202652.1 | - | 2 - 3 | 220 | 98 | ||
| AK204611.1 | - | 2 - 3 | 220 | 98 | ||
| AK205182.1 | - | 2 - 3 | 220 | 98 | ||
| BC010319.1 | - | 16 - 17 | 2451 | 98 | ||
| BC021496.1 | - | 18 - 19 | 2816 | 5346159 | 98 | |
| BC023175.1 | - | 2 - 3 | 160 | 98 | ||
| BC023794.1 | - | 19 - 20 | 2761 | 5351943 | 98 | |
| BC023867.1 | - | 1 - 2 | 162 | 98 | ||
| BC023964.1 | - | 18 - 19 | 2807 | 98 | ||
| BC025037.1 | - | 11 - 12 | 1666 | 98 | ||
| BC025618.1 | - | 18 - 19 | 2825 | 5320976 | 98 | |
| BC025627.1 | - | 18 - 19 | 2816 | 5320513 | 98 | |
| BC025811.1 | - | 18 - 19 | 2803 | 5096872 | 98 | |
| BC030326.1 | - | 7 - 8 | 1163 | 98 | ||
| BC031176.1 | - | 5 - 6 | 724 | 98 | ||
| BC031389.1 | - | 2 - 3 | 285 | 98 | ||
| BC032187.1 | - | 18 - 19 | 2866 | 5321268 | 98 | |
| BC033435.1 | - | 18 - 19 | 2762 | 4950746 | 98 | |
| BC033471.1 | - | 18 - 19 | 2806 | 4954119 | 98 | |
| BC034177.1 | - | 17 - 18 | 2571 | 98 | ||
| BC035352.1 | - | 17 - 18 | 2536 | 98 | ||
| BC042435.1 | - | 18 - 19 | 2816 | 4951288 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a1
Genomic Map