Assay Details
| Gene Symbol: Ptbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144904.2 | NP_659153.2 | 8 - 9 | 1094 | 72 | |
| NM_178164.3 | NP_835458.1 | 7 - 8 | 1069 | 72 | ||
| GenBank mRNA | AK033732.1 | - | 7 - 8 | 994 | 72 | |
| AK042920.1 | - | 5 - 6 | 1001 | 72 | ||
| AK088560.1 | - | 7 - 8 | 1001 | 72 | ||
| BC006638.1 | - | 8 - 9 | 1004 | 3969488 | 72 | |
| BC057641.1 | - | 7 - 8 | 974 | 4936557 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptbp3
Genomic Map